Print…
ratiolab® Pipette Tips, Colourless
Fornecedor: RATIOLAB
2400642
RATI2400642EA 29.9 EUR
RATI2400642
ratiolab® Pipette Tips, Colourless
Pontas de pipetas
ratiolab® pipette tips are suitable for all commonly used pipette types.
  • Precision in each and every detail
  • Guarantee for highest quality
  • Made of high quality polypropylene
  • Autoclavable
  • Excellent transparency due to high quality surfaces