Print…

RNA isolation, viral RNA kit, E.Z.N.A.®

Fornecedor: OMEGA BIO-TEK

R6874-02 R6874-01 R6874-00
R6874-02EA 714 EUR
R6874-02 R6874-01 R6874-00
RNA isolation, viral RNA kit, E.Z.N.A.®
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents

The E.Z.N.A.® Viral RNA Kit is designed for the isolation of viral RNA from cell free fluids such as plasma, serum, urine and cell culture supernatant. The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues and bacteria. RNA purified using the E.Z.N.A.® Viral RNA method is ready for all downstream applications such as RT-PCR.

Encomendar

ESPECIFICAÇÕES

Descrição E.Z.N.A.® Viral RNA Kit
Environmentally Preferable
Aplicação
Target molecule RNA
Refine target molecule Viral RNA
Sample type Biofluids (serum, plasma, saliva)
Method / Format Column (silica-based)
Prep size Mini
Tamanho da amostra 150 µl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR