Print…
USB Rechargeable Head Light
Fornecedor: Portwest
PA71BKR
PRTWPA71BKREA 14.4 EUR
PRTWPA71BKR
USB Rechargeable Head Light
Luzes Lanternas
Super bright COB head torch with 180° wide beam produces bright, even light. Ideal for night reading, car repair, or other close-up lighting applications.
  • Brightness: 100 lumens
  • Run time: 1 h (high), 5 h (low)
  • Recharge time: 4 h
  • Functions: high-low-red-red flash
  • Beam distance: 50 m
  • Retail box aids presentation for retail sales
Head light with tilt control allows aiming the light where it is needed most. Ideal for keeping hands free for work.
Fornecimento: Micro USB charging cable included.