Print…
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Fornecedor: PROTIDE PHARMACEUTICALS
PP338DM5-10 PP338DM5-50 PP338DM5-100 PP338DM5-100B PP338DM10-10 PP338DM10-50 PP338DM10-100 PP338DM10-100B 338DM10-1000B PP338DM20-100B 338DM20-1000B
PRPHPP338DM5-10EA 0 EUR
PRPHPP338DM5-10 PRPHPP338DM5-50 PRPHPP338DM5-100 PRPHPP338DM5-100B PRPHPP338DM10-10 PRPHPP338DM10-50 PRPHPP338DM10-100 PRPHPP338DM10-100B PRPH338DM10-1000B PRPHPP338DM20-100B PRPH338DM20-1000B
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Cultura celular, Suplementos e aditivos
When combined with DMSO, 2-8CELLsius™ can be utilised as a cryopreservation solution. 2-8CELLsius™ + DMSO is a cytoprotective, protein-free solution, with bio-stabilisers that support high post thaw cell viability. Optimal recovery is achieved through a novel approach to cell physiology at ultra-low temperatures.
  • TYPE II drug master file
  • USP grade components/highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones, antibiotics
  • Ready to use/sterile membrane filtered
Certificações: cGMP manufactured.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Endotoxinas <0,5 EU/ml at 1X
Espécies
Origem
Shelf life 20 months from DOM
Temperatura de armazenamento 2...8 °C