Print…

Ampicillin

Fornecedor: LGC Standards PROMOCHEM

MM0253.16
PROOMM0253.16EA 1990 EUR
PROOMM0253.16
Ampicillin
Ampicillin
Fórmula: C₁₆H₁₉N₃O₄S
Peso Molecular: 349.4 g/mol
Número MDL : MFCD00005175
Número CAS: 69-53-4
EINECS: 200-709-7

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR