Print…

2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202) 35 µg/ml in isooctane

Fornecedor: LGC Standards PROMOCHEM

CIL-PCB-202
PROOCIL-PCB-202EA 251 EUR
PROOCIL-PCB-202
2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202) 35 µg/ml in isooctane
Padrões Organic Standards

Encomendar

ESPECIFICAÇÕES

Descrição 2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202)
Environmentally Preferable
CAS No. 2136-99-4
Formula C₁₂H₂Cl₈

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR