Print…

Dibenzo[a,j]antraceno

Fornecedor: LGC Standards PROMOCHEM

BCR-095
PROOBCR-095EA 966 EUR
PROOBCR-095
Dibenzo[a,j]antraceno
Dibenzo[a,j]antraceno
Fórmula: C₂₂H₁₄
Peso Molecular: 278.35 g/mol
Número CAS: 224-41-9
EINECS: 205-928-1

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR