Print…

Restriction enzymes, Mbo I

Fornecedor: Promega

R6711
PROMR67110200EA 0 EUR
PROMR67110200
Restriction enzymes, Mbo I
Reagentes de ácidos nucleicos Restriction Enzymes

These Mbo I restriction enzyme recognises ˅GATC and CTAG^ sites and cuts best at 37 °C in its own unique buffer.


  • Sensitive to dam methylation
  • DpnI is an unusual isoschizomer of MboI that will cut only if the recognition site is methylated by the dam methylation system
  • Sau3AI is an isoschizomer of MboI that is not sensitive to dam methylation
  • Source: Moraxella bovis ATCC 10900


Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.


Fornecimento: Kit includes (1×200 U) Mbo I, (1×1 ml) buffer C 10X, (1×150 µl) bovine serum albumin (acetylated).

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR