Print…

Restriction enzymes, ApaI, HC

Fornecedor: Promega

R4364
PROMR436425KUEA 0 EUR
PROMR436425KU
Restriction enzymes, ApaI, HC
Reagentes de ácidos nucleicos Restriction Enzymes

These ApaI restriction enzyme recognises and cut G GGCC˅C and C^CCGG G at 37 °C in its own unique buffer.


  • Active and capable of digestion directly in GoTaq® Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Highly concentrated


Storage buffer: 10 mM Tris-HCl (pH 7,4), 300 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.


Source: Acetobacter pasteurianus.


Fornecimento: Kit includes 1×1ml buffer A 10X buffer, 1×750 µl bovine serum albumin, acetylated with concentration of 10 mg/ml , 1×5000 units ApaI and 1×0,25 ml MULTI-CORE™ 10X buffer.

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR