Print…

Brometo de etídio 10 mg/ml aqueous solution Molecular_Biology

Fornecedor: Promega
Atenção

Sinônimos: Brometo de 3,8-diamino-5-etil-6-fenilfenantridínio

H5041
PROMH50410010EA 87.2 EUR
PROMH50410010
Brometo de etídio 10 mg/ml aqueous solution Molecular_Biology
Brometo de etídio

Ethidium Bromide Solution, Molecular Grade (10mg/ml), is a fluorescent dye suitable for staining nucleic acids after electrophoresis or in cesium chloride gradients. The solution can be used to detect both double-stranded and single-stranded DNA.

Fórmula: C₂₁H₂₀BrN₃
Peso Molecular: 394.31 g/mol
Ponto de ebulição: 72
Número MDL : MFCD00011724
Número CAS: 1239-45-8
EINECS: 214-984-6
Merck Index: 13,04751

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR