Print…
Dual-Luciferase® reporter (DLR™) assay system
Fornecedor: Promega
E1910 E1941 E1960 E1980
PROME19100100EA 433 EUR
PROME19100100 PROME19410030 PROME19600010 PROME19801000
Dual-Luciferase® reporter (DLR™) assay system
Ensaios Ensaios celulares
The Dual-Luciferase® Reporter (DLR™) Assay System provides an efficient means of performing two reporter assays. In the DLR™ Assay, the activities of firefly (Photinus pyralis) and Renilla (Renilla reniformis or sea pansy) luciferases are measured sequentially from a single sample.
  • Renilla luciferase internal control provides more accurate results
  • Range extends over 7 logs; very active samples typically do not need dilution
  • Requires luminometer with 2 injectors
The pGL4 Luciferase Reporter Vectors are designed for use with the DLR™ Assay Systems. A Renilla luciferase vector with constitutive expression may be used in combination with any experimental firefly luciferase vector to co-transfect mammalian cells.
Fornecimento: For applications requiring more lysis reagent (e.g., >100 µl/well), additional Passive Lysis Buffer may be purchased separately.