Print…
Cassette printer
Fornecedor: Primera Technology Europe
078408
PRMA078408EA 18280 EUR
PRMA078408
Cassette printer
Impressoras Cassette and Slide Printers
Cassette printer reduces the risk of misidentification of specimens. It helps to print text, graphics and logos along with linear and 2D bar codes directly onto the face of the cassettes with eight solid colours.
  • Chemical resistant ink
  • Steel frame with powder coated steel covers
  • Ribbon types: CMYK (550 prints) and black (4300 prints)
Printed speed: 8 cassettes per minute, 4 cassette hoppers and 40 cassettes per hopper.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Alimentação 50/60 Hz, 60 W
Interfaces USB 2.0 compatible
Resolução 300 dpi
Supported OS Windows™ 7/10