Print…
DAB substrate kit, metal enhanced, Pierce™
Fornecedor: Thermo Fisher Scientific
34065
PIER34065EA 298 EUR
PIER34065
DAB substrate kit, metal enhanced, Pierce™
Electroforese, Reagentes Western Blotting Reagents
The Pierce™ Metal enhanced DAB substrate kit uses cobalt and nickel chloride in a special formulation of diaminobenzidine peroxidase substrate that yields intense colour for immunohistochemical staining.
  • Up to 50 times more sensitive than traditional DAB methods
  • Low background and high intensity
  • Only 2 components
  • Working solution stable for more than six hours at room temperature