Print…
Substrate buffers, Pierce™
Fornecedor: Thermo Fisher Scientific
34064 34062
PIER34064EA 98.75 EUR
PIER34064 PIER34062
Substrate buffers, Pierce™
Tampões Tampões enzimáticos
These stable peroxide and diethanolamine substrate buffers produce optimised, reliable and convenient stock solutions for HRP and alkaline phosphatase assays detection respectively.
  • Ready to use
  • Minimises assay-to-assay variability