Print…
Pharmaceutical secondary standard, L-Glutamic acid, CRM, Supelco®
Fornecedor: Merck
PHR1107-1G
PHR1107-1GEA 156 EUR
PHR1107-1G
Pharmaceutical secondary standard, L-Glutamic acid, CRM, Supelco®
Padrões Organic Standards
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
These secondary standards offer multi-traceability to the USP, EP (Ph. Eur.) and BP primary standards, where they are available.
Certificações: Accordance with ISO/IEC 17025:2005 (A Class Cert AT-1467), ISO GUIDES 34:2009 (A Class Cert AR-1470).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5293898d25f5ab4efa0e93f67d6e5f2b53a33e70&vid=5293898d25f5ab4efa0e93f67d6e5f2b53a33e70&prid=web&clid=SAPDAM