Print…
Racks for SPB system tubes
Fornecedor: PerkinElmer
N9308070
PERCN9308070EA 314 EUR
PERCN9308070
Racks for SPB system tubes
Suportes/Racks Tube Racks
Polycarbonate.
  • With rack lock feature which locks DigiTUBE®s in position within the rack allowing for easy and one-handed closure of screw cap
  • Acid resistant
  • For laboratory usage