Print…
Polarimeter cells
Fornecedor: PerkinElmer
B0017054
PERCB0017054EA 2210 EUR
PERCB0017054
Polarimeter cells
Cuvetes Cuvetes especiais
Glass cells designed for easy filling and emptying with no loss of sample.
  • High efficiency, thermostattable, easy to fill and empty with no loss of sample
  • Filling ports are arranged so that the liquid flows over the inner window surfaces
  • Windows are fused to the cell bodies to ensure tension free operation
  • Defined pathlength engraved on each cell for easy identification
  • Cells are jacketed for thermostatting with constant temperature water, except the flowcell