Print…
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Fornecedor: Avantor Fluid Handling
PAWBEVSS-CFLEX0.5EA 1240 EUR
PAWBEVSS-CFLEX0.5 PAWBEVSS-CFLEX0.75 PAWBEVSS-CFLEX1.0 PAWBEVSS-CFLEX1.25 PAWBEVSS-CFLEX1.5 PAWBEVSS-CFLEX1.75 PAWBEVSS-CFLEX10.1 PAWBEVSSCFLEX10.24 PAWBEVSS-CFLEX10.4 PAWBEVSSCFLEX10.74 PAWBEVSS-CFLEX11.1 PAWBEVSS-CFLEX2.0 PAWBEVSS-CFLEX2.25 PAWBEVSS-CFLEX2.5 PAWBEVSS-CFLEX2.75 PAWBEVSS-CFLEX3.0 PAWBEVSS-CFLEX3.25 PAWBEVSS-CFLEX3.5 PAWBEVSS-CFLEX3.75 PAWBEVSS-CFLEX4.0 PAWBEVSS-CFLEX4.25 PAWBEVSS-CFLEX4.5 PAWBEVSS-CFLEX4.75 PAWBEVSS-CFLEX5.0 PAWBEVSS-CFLEX5.25 PAWBEVSS-CFLEX5.5 PAWBEVSS-CFLEX5.75 PAWBEVSS-CFLEX6.0 PAWBEVSS-CFLEX6.25 PAWBEVSS-CFLEX6.5 PAWBEVSS-CFLEX6.75 PAWBEVSS-CFLEX7.0 PAWBEVSS-CFLEX7.25 PAWBEVSS-CFLEX7.5 PAWBEVSS-CFLEX7.75 PAWBEVSS-CFLEX8.0 PAWBEVSS-CFLEX8.25 PAWBEVSS-CFLEX8.5 PAWBEVSS-CFLEX8.75 PAWBEVSS-CFLEX9.0 PAWBEVSS-CFLEX9.25 PAWBEVSS-CFLEX9.5 PAWBEVSS-CFLEX9.75 PAWBEVSS-PVC0.5ML PAWBEVSS-PVC0.74 PAWBEVSS-PVC1.0ML PAWBEVSS-PVC1.24 PAWBEVSS-PVC1.5ML PAWBEVSS-PVC1.74 PAWBEVSS-PVC10.0 PAWBEVSS-PVC10.25 PAWBEVSS-PVC10.5 PAWBEVSS-PVC10.75 PAWBEVSS-PVC11.1 PAWBEVSS-PVC2.0ML PAWBEVSS-PVC2.24 PAWBEVSS-PVC2.5ML PAWBEVSS-PVC2.74 PAWBEVSS-PVC3.0ML PAWBEVSS-PVC3.24 PAWBEVSS-PVC3.5ML PAWBEVSS-PVC3.74 PAWBEVSS-PVC4.0ML PAWBEVSS-PVC4.24 PAWBEVSS-PVC4.5ML PAWBEVSS-PVC4.74 PAWBEVSS-PVC5.0ML PAWBEVSS-PVC5.24 PAWBEVSS-PVC5.5ML PAWBEVSS-PVC5.74 PAWBEVSS-PVC6.0ML PAWBEVSS-PVC6.24 PAWBEVSS-PVC6.5ML PAWBEVSS-PVC6.74 PAWBEVSS-PVC7.0ML PAWBEVSS-PVC7.24 PAWBEVSS-PVC7.5ML PAWBEVSS-PVC7.74 PAWBEVSS-PVC8.0ML PAWBEVSS-PVC8.24 PAWBEVSS-PVC8.5ML PAWBEVSS-PVC8.74 PAWBEVSS-PVC9.0ML PAWBEVSS-PVC9.24 PAWBEVSS-PVC9.5ML PAWBEVSS-PVC9.74
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Tubos Tubos para ensaio
Optimise sampling processes with the Exact Volume Sampling Systems (EVSS) from OmniTop Sample Tubes®.
  • Customised sample volume at point of use
  • Aseptic, closed system sampling reduces contamination risk
  • Improved product yield
  • Reduced assembly and installation time
  • Easy operation and closed system transfer
Choose any volume between 0,5 to 11,0 ml and take exact volume samples with a ±0,25 ml accuracy. The EVSS is available with a C-Flex® tubing inlet or a PVC (Tygon®) tubing option with a male Luer end connection.
The EVSS can be asepticallyMais informação do produto