Print…
Cleanroom tables, Ergo
Fornecedor: Palbam
WX0901 WX0901CASTORS WX0902 WX0902CASTORS WX0903 WX0903CASTORS WX0904 WX0904CASTORS WX0905 WX0905CASTORS WX0906 WX0906CASTORS WX1201CASTORS WX1202 WX1202CASTORS WX1204 WX1204CASTORS WX1205 WX1205CASTORS WX1206 WX1206CASTORS WX1401CASTORS WX1402 WX1402CASTORS WX1403CASTORS WX1404 WX1404CASTORS WX1405 WX1405CASTORS WX1406 WX1406CASTORS WX0804 WX2004 WX1602 WX0802 WX1504 WX1554 WX2002 WX1253C WX0951C WX0601 WX1251 WX1453C WX1553C WX1001 WX0702 WX1051 WX0952C WX0651 WX2001 WX1751 WX0953-C WX0801 WX1502 WX2051 WX2052
132-0300EA 2100 EUR
132-0300 132-0301 132-0306 132-0307 132-0312 132-0313 132-0318 132-0319 132-0324 132-0325 132-0330 132-0331 132-0303 132-0308 132-0309 132-0320 132-0321 132-0326 132-0327 132-0332 132-0333 132-0305 132-0310 132-0311 132-0317 132-0322 132-0323 132-0328 132-0329 132-0334 132-0335 132-1062 132-1075 PALBWX1602 PALBWX0802 PALBWX1504 PALBWX1554 PALBWX2002 PALBWX1253C PALBWX0951C PALBWX0601 PALBWX1251 PALBWX1453C PALBWX1553C PALBWX1001 PALBWX0702 PALBWX1051 PALBWX0952C PALBWX0651 PALBWX2001 PALBWX1751 PALBWX0953-C PALBWX0801 PALBWX1502 PALBWX2051 PALBWX2052
Cleanroom tables, Ergo
Mobiliário Mesas
These cleanroom tables are manufactured to the highest cleanroom standards. The bases are fabricated from tubing for easy cleaning and optimal airflow.
  • Tables have fixtures enabling the accessories to be attached either at the time of purchase or in the future if the need arises
  • The tops are designed to reduce vibrations and are very strong supporting loads up to 80 kg
Stainless steel 304 with electropolished finish and anodised aluminium connectors.
Fornecimento: The tables are shipped flat-packed with easy assembly instructions.