Print…
Avantor® Partisil SAX, HPLC Columns
Fornecedor: Avantor
P10SAX-250AEA 1150 EUR
P10SAX-250A P5SAX-250A P10SAX-10C5 P10SAX-150A P10SAX-150AS P10SAX-250AF P5SAX-100A P5SAX-10C5 P5SAX-10CM5 P5SAX-250AF P5SAX-250AS P10SAX-100P P10SAX-10C5-SK HICHP5SAX-10CM5-SK
Avantor® Partisil SAX, HPLC Columns
Cromatografia, Colunas
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® SAX is a strong anion exchange phase based on quaternary ammonium groups and is stable over the pH range 1,5 to 7,5. At intermediate pH, use of a Solvecon eluent conditioning column is recommended to further enhance column lifetime. Suitable for use with nucleic acids, organic acids and inorganic anions.
Atenção: * Use with freestanding holder HI-161 andMais informação do produto