Print…
Avantor® Partisil Silica, HPLC Columns
Fornecedor: Avantor
P5-250AEA 1220 EUR
P5-250A P10-250A P5-150A P5-200A P5-250AS P10-150AF P10-250AF P5-100A P5-250AF
Avantor® Partisil Silica, HPLC Columns
Cromatografia, Colunas
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity. Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.
  • Irregular porous silica
  • 5 and 10 µm particle sizes
  • Hichrom high efficiency