Print…
Avantor® Partisil SCX, HPLC Columns
Fornecedor: Avantor
P10SCX-100AEA 1130 EUR
P10SCX-100A P10SCX-250A P10SCX-10C5 P5SCX-250A P5SCX-125A P5SCX-150A P10SCX-150A P10SCX-250AF P5SCX-100A P5SCX-100AM P5SCX-10C5 P5SCX-10CM5 P5SCX-250AF P5SCX-50A P10SCX-10C5-SK
Avantor® Partisil SCX, HPLC Columns
Cromatografia, Colunas
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® SCX is a strong cation exchange phase based on benzenesulphonic acid groups and is stable over the pH range 1,5 to 7,0. At intermediate pH, use of a Solvecon eluent conditioning column is recommended to further enhance column lifetime. Suitable for use with nucleosides, amino acids, polyamines, drugs and other cationic species.
Atenção: * Use withMais informação do produto