Print…
Culti-Loops™ Fusobacterium necrophorum ATCC™ 25286™
Fornecedor: THERMO FISHER DIAGNOSTICS
R4609375
OXOIR4609375EA 262.5 EUR
OXOIR4609375
Culti-Loops™ Fusobacterium necrophorum ATCC™ 25286™
Testes para microbiologia Testes de Microrganismos
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Embalagem: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.