Print…
Blood culture system, Signal®
Fornecedor: THERMO FISHER DIAGNOSTICS
BC0100M BC0104A
OXOIBC0100MEA 305 EUR
OXOIBC0100M OXOIBC0104A
Blood culture system, Signal®
Sistemas para diagnóstico Clinical Sample Collection/Preparation Coleta/Preparação de Amostras de Sangue
Blood samples are collected from patients, using strict aseptic technique and sterile equipment. The samples are inoculated into the blood culture bottles and mixed with the medium.
  • Well-proven
  • Reliable
The formulation of the medium encourages the growth of aerobic, anaerobic and micro-aerophilic organisms. The medium is also designed to create pressure in the sealed bottle when organisms are growing. The detection of positive pressure is by means of a growth indicator device which is connected to the bottle after the blood sample is added.