Print…
Precision tweezers, resistant, DURAX™
Fornecedor: OUTILS RUBIS
1-DURAX 00-DURAX 0-DURAX 2A-DURAX 3C-DURAX 3-DURAX 5A-DURAX 5-DURAX 7-DURAX SS-DURAX
OUTI1-DURAXEA 24.2 EUR
OUTI1-DURAX OUTI00-DURAX OUTI0-DURAX OUTI2A-DURAX OUTI3C-DURAX OUTI3-DURAX OUTI5A-DURAX OUTI5-DURAX OUTI7-DURAX OUTISS-DURAX
Precision tweezers, resistant, DURAX™
Forceps
DURAX™ (Martensitic steel).
  • Hardened up to 616 Vickers: becomes six times harder than standard steel
  • High resistance against wear and tear: Longer service life than ordinary tweezers