Print…

Human Recombinant Ikaros (IKZF1) (NM_006060)

Fornecedor: OriGene

TP313207M
ORIGTP313207MEA 4100 EUR
ORIGTP313207M
Human Recombinant Ikaros (IKZF1) (NM_006060)
Proteínas e Peptídeos

This gene encodes a transcription factor that belongs to the family of zinc-finger DNA-binding proteins associated with chromatin remodeling. The expression of this protein is restricted to the fetal and adult hemo-lymphopoietic system, and it functions as a regulator of lymphocyte differentiation.

Novo produto

Encomendar

ESPECIFICAÇÕES

Protein/peptide name Ikaros (IKZF1)
Protein synonyms PRO0758|ZNFN1A1|LyF-1|PPP1R92|CVID13|LYF1|IK1|IKAROS|Hs.54452
Environmentally Preferable
Protein/peptide type Recombinante
Espécies Human
Origem
UniProtKB Q13422, R9R4D9
Purity >80% as determined by SDS-PAGE and Coomassie blue staining
Concentração >0,05 µg/µl as determined by microplate BCA method
Molecular weight 57,3 kDa
Storage conditions 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol, Store at –80 °C
Tag sequence C-Myc/DDK

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR