Print…

Anti-COL15A1 Mouse Monoclonal Antibody [clone: OTI1C5]

Fornecedor: OriGene
TA507124
ORIGTA507124EA 621 EUR
ORIGTA507124
Anti-COL15A1 Mouse Monoclonal Antibody [clone: OTI1C5]
Anticorpos
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: COL15A1 (collagen, type XV, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR