Print…

Anti-IDH1 Mouse Monoclonal Antibody [clone: OTI2H9]

Fornecedor: OriGene
TA500610
ORIGTA500610EA 621 EUR
ORIGTA500610
Anti-IDH1 Mouse Monoclonal Antibody [clone: OTI2H9]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: IDH1 (isocitrate dehydrogenase 1 (NADP+), soluble)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR