Print…

Anti-TMEM100 Mouse Monoclonal Antibody [clone: OTI1F8]

Fornecedor: OriGene
TA500508
ORIGTA500508EA 621 EUR
ORIGTA500508
Anti-TMEM100 Mouse Monoclonal Antibody [clone: OTI1F8]
Anticorpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: TMEM100 (transmembrane protein 100)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Encomendar

ESPECIFICAÇÕES

Símbolo do antígeno TMEM100
Nome do antigénio transmembrane Protein 100
Environmentally Preferable
Tipo de anticorpo Primário
Clonalidade Monoclonal
Conjugado Não conjugado
Clone OTI1F8
Reatividade
Hospedeiro Mouse
Isótipo IgG2b
Western blot Yes
ImunoQuímica Yes
ImunoFluorescência Yes
Epítopo Full length
Forma Liquid
Molecular weight 14.4 kDa
Purification purified by affinity chromatography
Adsorção cruzada none
Storage buffer PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Temperatura de armazenamento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR