Print…

Anti-HLA-G Mouse Polyclonal Antibody [clone: 87G]

Fornecedor: OriGene
TA320347
ORIGTA320347EA 299 EUR
ORIGTA320347
Anti-HLA-G Mouse Polyclonal Antibody [clone: 87G]
Anticorpos
Type: Primary
Antigen: HLA-G
Clonality: Polyclonal
Clone: 87G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR