Print…

Anti-ITCH Goat Polyclonal Antibody

Fornecedor: OriGene
TA302519
ORIGTA302519EA 785 EUR
ORIGTA302519
Anti-ITCH Goat Polyclonal Antibody
Anticorpos
Recommended Dilutions: ELISA: 1:32000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: ITCH (itchy E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Encomendar

ESPECIFICAÇÕES

Símbolo do antígeno ITCH
Environmentally Preferable
Nome do antigénio itchy E3 ubiquitin Protein ligase
Tipo de anticorpo Primário
Clonalidade Policlonal
Conjugado Não conjugado
Reatividade Human,Dog,Rat,Mouse
Hospedeiro Goat
Isótipo IgG
Western blot Yes
ELISA Yes
ImunoQuímica Yes
Epítopo internal
Sequence C-EIKSHDLKPNGGN
Forma Liquid
Molecular weight 102.31 kDa
Purification purified by ammonium sulphate precipitation followed by antigen affinity chromatography
Adsorção cruzada none
Storage buffer Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
Temperatura de armazenamento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR