Print…

Anti-ING2 Mouse Monoclonal Antibody [clone: 4G10]

Fornecedor: OriGene
CF800665
ORIGCF800665EA 833 EUR
ORIGCF800665
Anti-ING2 Mouse Monoclonal Antibody [clone: 4G10]
Anticorpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: ING2
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR