Print…

Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1C2]

Fornecedor: OriGene
CF504862
ORIGCF504862EA 833 EUR
ORIGCF504862
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1C2]
Anticorpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:50

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 1C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR