Print…

Anti-ACBD3 Mouse Monoclonal Antibody [clone: 2B12]

Fornecedor: OriGene
CF504857
ORIGCF504857EA 833 EUR
ORIGCF504857
Anti-ACBD3 Mouse Monoclonal Antibody [clone: 2B12]
Anticorpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ACBD3
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR