Print…
Wall-mount mat roll dispensers, PIG®
Fornecedor: NEW PIG
MAT292
NEWPMAT292EA 124 EUR
NEWPMAT292
Wall-mount mat roll dispensers, PIG®
Dispensadores para Toalhetes e Panos
These mat roll dispensers feature a braking mechanism for more controlled dispensing.
  • Easy to refill and replace
  • Slot and cork brake prevents mat from unfurling as it's dispensed
  • Durable hot-rolled steel frame and galvanised core rod built to last in industrial settings
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable