Print…
PIG® Fat Mat® super-absorbent mat
Fornecedor: NEW PIG
MAT2101
NEWPMAT2101EA 135.5 EUR
NEWPMAT2101
PIG® Fat Mat® super-absorbent mat
Materiais para controlo de derrames
This double-thick version of 8-layer PIG Mat has 16 thirsty layers for big cleanups. Spunbond top resists abrasion for long life.
  • 16 layers of 100% polypropylene are thermally bonded to form one super-absorbent mat capable of drinking up huge volumes of liquid
  • Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated
  • Fast-wicking 'diaper action' transfers liquids into the absorbent inner layers to keep moisture away from surface
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-
Mais informação do produto