Print…
PIG® Burpless® Poly Drum Funnel
Fornecedor: NEW PIG
DRM138-RD DRM138-YW
NEWPDRM138-RDEA 120.5 EUR
NEWPDRM138-RD NEWPDRM138-YW
PIG® Burpless® Poly Drum Funnel
Funis Funis de segurança
Burpless design vents during pouring for fast, smooth flow with no dangerous splashback.
  • Lets you pour without venting through 3/4" bung
  • Thick-wall, 100% polyethylene construction resists UV rays, rust, corrosion and most chemicals
  • Designed for use with non-hazardous liquids: coolants, non-flammable oils and water-based liquids
  • Lockable to prevent unauthorised access
  • Multiple lid colour options let you segregate drums
  • Attached hinged lid won't get lost or separated from funnel
  • Ridged interior creates a level draining surface for containers and promotes
Mais informação do produto