Print…
Jerricãs, planos, com torneira e tampa roscada, Lowboys™, Nalgene®
Fornecedor: Thermo Fisher Scientific
2324-0015
NALG23240015EA 379 EUR
NALG23240015
Jerricãs, planos, com torneira e tampa roscada, Lowboys™, Nalgene®
Frascos Jerricans
Made of PP, translucent.
  • Can be stacked three high
  • Autoclave between uses to eliminate bacterial contamination
  • Convenient integral handgrip for safe, easy transport
Carboys have a low-profile design which fits easily into tight spaces, on shelves or in lab refrigerators. Ideal for storing and dispensing reagents.