Print…
Spray adhesive, Hi-Strength 90
Fornecedor: 3M
90
MMMA90EA 294 EUR
MMMA90
Spray adhesive, Hi-Strength 90
Adesivos
3M™ hi-strength 90 spray adhesive is an extremely versatile, fast-drying spray adhesive that bonds strongly to a wide range of materials. A permanent bond ensures fast results to keep projects moving ahead.
  • Provides professional, industrial strength adhesion for demanding applications
  • Creates soft, non-dimpling adhesive lines for a clean appearance
  • High temperature and moisture resistance
  • Variable-width spray pattern for more control and minimum waste
The spray formula is easy to dispense, offering a variable-width spray pattern, and applies directlyMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable