Print…

Shoe covers, 442

Fornecedor: 3M
442
MMMA442EA 196 EUR
MMMA442
Shoe covers, 442
Roupas Descartáveis
These disposable shoe covers are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material (PP/PE) with anti-static treatment.

  • Elastic around the ankle for protection against contaminants and enhanced fit
  • Sole and upper constructed of the same material
  • Anti-static treated

Weight: 55 g/m²

Length: 380 mm
Height: 180 mm

Opening diameter: 8 - 15 cm
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR