Print…
Microplate readers, multi-mode, SpectraMax® M series
Fornecedor: MOLECULAR DEVICES
M2 M2-SC-ACAD M2E M2E-SC-ACAD M3 M3-SC-ACAD M4 M4-SC-ACAD M5 M5-SC-ACAD M5E M5E-SC-ACAD
MLDVM2EA 0 EUR
MLDVM2 MLDVM2-SC-ACAD MLDVM2E MLDVM2E-SC-ACAD 736-0910 MLDVM3-SC-ACAD MLDVM4 MLDVM4-SC-ACAD MLDVM5 MLDVM5-SC-ACAD MLDVM5E MLDVM5E-SC-ACAD
Microplate readers, multi-mode, SpectraMax® M series
Leitores de microplacas
All SpectraMax® M series multi-mode microplate readers measure UV and visible absorbance, and fluorescence intensity measurements on microplates and cuvettes as standard. The SpectraMax® M2 and M2e readers provide two detection modes in one platform, and offer the the added functionality of bottom-read fluorescence for cell-based assays (M2e). The SpectraMax® M3, M4, M5 and M5e readers offer a modular, upgradeable dual monochromator microplate reader platform with a wide range of high performance multi-mode capabilities ideal for life science research and drug discovery screening.