Print…
Millex Samplicity® Filters
Fornecedor: Merck
SAMPHV001 SAMPLCR01 SAMPLG001 SAMPLCR04 SAMPLG004 SAMPHV004
MILFSAMPHV001EA 445 EUR
MILFSAMPHV001 MILFSAMPLCR01 MILFSAMPLG001 MILFSAMPLCR04 MILFSAMPLG004 MILFSAMPHV004
Millex Samplicity® Filters
Filtros Filtros sem Seringa
Millex Samplicity® Filters for the Samplicity® filtration system.
  • Samplicity® Filtration System enables vacuum driven filtration of up to eight samples at once, directly into HPLC vials
  • Filtration for HPLC sample prep without syringes
  • Millex Samplicity® Filters provide integrated funnels and filters for easiest sample loading
  • Available in 0,20 µm and 0,45 µm pore sizes with hydrophilic PTFE and Durapore® PVDF membranes
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable