Print…
Samplicity® G2 Filtration System and Filters
Fornecedor: Merck
SAMP2SYSB SAMP2SYSG SAMP2GVNB SAMP2HVNB SAMP2GNNB SAMP2HNNB SAMP2HPNB
MILFSAMP2SYSBEA 1850 EUR
MILFSAMP2SYSB MILFSAMP2SYSG MILFSAMP2GVNB MILFSAMP2HVNB MILFSAMP2GNNB MILFSAMP2HNNB MILFSAMP2HPNB
Samplicity® G2 Filtration System and Filters
Filtros Filtros sem Seringa
Vacuum filtration system for sample preparation for HPLC. Samplicity® G2 filtration system enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Minimal hold-up volume
  • Filter and seal integrity
  • Low extractables, low binding
  • Integrated funnels for easiest sample loading
  • Filters in perforated strips of 4 for fastest setup
  • Lower ergonomic impact than manual syringe filtration
  • Non sterile
The adapter funnels provided for the Samplicity® G2 system are made of the same polypropylene as our Millex® filter housings.
Mais informação do produto