Print…
Masterflex® I/P® Precision Pump Tubing, Puri-Flex®, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX96419-26EA 177 EUR
MFLX96419-26 MFLX96419-73 MFLX96419-82
Masterflex® I/P® Precision Pump Tubing, Puri-Flex®, Avantor®
Tubagem Tubulação de Bomba Peristáltica
High-performance and long-life tubing for high-purity applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Low spallation when compared to silicone or some