Print…
Masterflex® Transfer Tubing Spool, Norprene®, Avantor®
Fornecedor: Avantor Fluid Handling
95901-08
MFLX95901-08EA 976 EUR
MFLX95901-08
Masterflex® Transfer Tubing Spool, Norprene®, Avantor®
Tubagem Tubulação de transferência
Resists chemicals and UV-light.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range isMais informação do produto