Print…
Masterflex® MasterSense™ Piston Pump Systems
Fornecedor: Avantor Fluid Handling
MFLX77976-60-EUEA 7850 EUR
MFLX77976-60-EU MFLX77976-61-EU MFLX77976-62-EU
Masterflex® MasterSense™ Piston Pump Systems
Bombas Bombas de pistão
Versatile piston pump systems for high-accuracy fluid delivery and dispensing with an intuitive touch-screen interface.
  • Powerful, space-efficient drives will accept FMI Q-Series and RH-Series piston pump heads
  • Piston pump technology offers high accuracy dispensing and repeatability
  • Q-Series FMI Pump heads feature fine tune stroke length adjustment for precision volume control
  • High pressure capability (up to 100 psi)
  • 12,7 cm (5'') capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Single-point pressure monitoring provides real-
Mais informação do produto