Print…
Masterflex® L/S® Precision Process Pump Systems, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX77916-42-EUEA 5270 EUR
MFLX77916-42-EU MFLX77916-40-EU MFLX77916-50-EU MFLX77916-52-EU
Masterflex® L/S® Precision Process Pump Systems, Avantor®
Bombas Bombas Peristálticas
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
  • Separate speed and power (on/off) control—
Mais informação do produto