Print…
Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX77913-60-EUEA 2850 EUR
MFLX77913-60-EU MFLX77913-70-EU MFLX77913-80-EU
Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
Bombas Bombas Peristálticas
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Choose between benchtop controller or washdown wall-mount controller drives
L/S® modular drives feature reversible pumping and allow you the flexibility to separate the motor from theMais informação do produto