Print…
Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX07019-00EA 764 EUR
MFLX07019-00 MFLX07019-25 MFLX07019-26 MFLX77601-62 MFLX77602-02 MFLX77602-12
Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Bombas Cabeças de bomba Cabeças de bomba peristáltica
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Designed and optimized for one tubing size
  • Three roller design for smooth flow
  • Choose from stainless steel (SS) or cold-rolled steel (CRS) rotor / roller for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
Standard heads are precision molded to accommodate a single size of tubing—use for common fluid transfer applications requiring accuracy and excellent repeatability. AvailableMais informação do produto