Print…
Micropump® B-Mount Gear Pump Heads
Fornecedor: Avantor Fluid Handling
74004-03
MFLX74004-01EA 1660 EUR
MFLX74004-01 MFLX74004-05 MFLX74004-07 MFLX74004-09 MFLX74004-11 MFLX74004-17 MFLX74004-23 MFLX74004-27 MFLX74004-29 MFLX74004-31 MFLX74004-33 MFLX74004-71 MFLX74004-73 MFLX74004-75 MFLX74004-03
Micropump® B-Mount Gear Pump Heads
Bombas Cabeças de bomba Cabeças de bomba de engrenagem
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads areMais informação do produto